Learn More
Corning™ PYREX™ 60° Angle Fluted Funnel, Short Stem
GREENER_CHOICE
Useful in clinical and hospital labs; can be used in flasks or tubes without racks
$339.10 - $505.03
Specifications
Material Borosilicate Glass
Angle 60°
For Use With (Application) Useful in clinical and hospital labs, can be used in flasks or tubes without racks
Product Type Fluted Filling Funnel
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric)Top Diameter (Metric) Stem Length (Metric) Stem Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric)Top Diameter (Metric) Stem Length (Metric) Stem Height (Metric) Price Quantity & Availability  
10329D
GREENER_CHOICE
View Documents Promotion Details
Corning™
6180100
10 cm 0.9 cm 10 cm 19 cm
Pack of 6 Each for $339.10
Case of 4 Pack for $1,356.40
Only null left
10329C
GREENER_CHOICE
View Documents Promotion Details
Corning™
618075
7.5 cm 0.8 cm 7.5 cm 13.9 cm
Pack of 12 Each for $416.50
Case of 4 Pack for $1,666.00
Only null left
10329A
GREENER_CHOICE
View Documents Promotion Details
Corning™
618050
5 cm 0.8 cm 6.5 cm 10.8 cm
Pack of 12 Each for $383.22
Case of 4 Pack for $1,532.88
Only null left
10329B
GREENER_CHOICE
View Documents Promotion Details
Corning™
618065
6.5 cm 0.8 cm 6.5 cm 12.1 cm
Pack of 12 Each for $505.03
Case of 4 Pack for $2,020.12
Only null left
Description
Description
Supplied with short stems. Depressed inside flutings further decreases filtering time.
Specifications
Specifications
Borosilicate Glass
60°
Useful in clinical and hospital labs, can be used in flasks or tubes without racks
Fluted Filling Funnel