Learn More
PYREX™ Enlarging Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Supplier: PYREX™ 88202429
Description
With an outer standard taper joint at the top and a larger inner standard taper joint at the bottom.
Specifications
| For attaching apparatus with one joint size to equipment having larger size joint | |
| 100 mm | |
| Tube Adapter |
| 3.93 in. | |
| Borosilicate Glass | |
| 29/42 Inner Bottom, 24/40 Outer Top |