missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Economy, Double Scale Cylinders

Borosilicate cylinders calibrated “To Contain” (TC)

$40.35 - $40.35

Specifications

Material Borosilicate Glass
Class B
For Use With (Application) Institutions
Scale Dual
Sterility Non-Sterile
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
S63259
View Documents
Fisher Science Education
302525EMD
25 mL 0.2 mL
Each for $40.35
Case of 18 Each for $726.30
 
Description

Description

 

  • Two metric scales for reading up or down
  • Shorter than standard cylinders
  • Without bumper guards

 

Specifications

Specifications

Borosilicate Glass
Institutions
Non-Sterile
B
Dual
SDS
Product Certifications