missing translation for 'onlineSavingsMsg'
Learn More

Pyrene 97.0+%, TCI America™

Catalog No. P110425G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P110425G 25 g
P1104100G 100 g
P1104500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. P110425G missing translation for 'mfr' TCI America missing translation for 'supplierNo' P110425G

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.256
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

Melting Point 150°C
Boiling Point 404°C
Quantity 25 g
UN Number 2811
Formula Weight 202.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrene
EINECSNumber (4)-0782
RTECSNumber UR2450000
TSCA Yes