Learn More
Pyrene, 98%
CAS: 129-00-0 | C16H10 | 202.256 g/mol
Supplier: Thermo Scientific Chemicals L0816222
Description
Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
| Pyrene | |
| 148°C to 152°C | |
| 393°C | |
| C16H10 | |
| 100 g | |
| 14,7963 | |
| Soluble in toluene. Insoluble in water. | |
| C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 | |
| 202.256 | |
| CHEBI:39106 | |
| 98% |
| 129-00-0 | |
| 1.271 | |
| 208°C (406°F) | |
| MFCD00004136 | |
| 1307225 | |
| benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
| BBEAQIROQSPTKN-UHFFFAOYSA-N | |
| pyrene | |
| 31423 | |
| 202.26 |
Chemical Identifiers
| 129-00-0 | |
| 202.256 | |
| BBEAQIROQSPTKN-UHFFFAOYSA-N | |
| 31423 | |
| pyrene |
| C16H10 | |
| MFCD00004136 | |
| benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
| CHEBI:39106 | |
| C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Safety and Handling
GHS H Statement
H332
Harmful if inhaled.
P260-P271-P304+P340-P312-P314-P501c
H336-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 204-927-3
RTECSNumber : UR2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only