missing translation for 'onlineSavingsMsg'
Learn More

Pyrene-1-boronic acid, 95%

Catalog No. AAH6421103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6421103 1 g
AAH6421106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6421103 Supplier Thermo Scientific Chemicals Supplier No. H6421103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 164461-18-1 | C16H11BO2 | 246.07 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field. Also used as a intermediate for organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 164461-18-1
Molecular Formula C16H11BO2
Molecular Weight (g/mol) 246.07
MDL Number MFCD04974062
InChI Key MWEKPLLMFXIZOC-UHFFFAOYSA-N
Synonym 1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid, boronic acid, 1-pyrenyl, 1-pyrene boronic acid, pyreneboronic acid, pubchem16490, pyren-1-yl boronic acid
PubChem CID 5084102
IUPAC Name pyren-1-ylboronic acid
SMILES OB(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 247°C to 251°C (literature)
Color Yellow
Quantity 1 g
Beilstein 7134073
Solubility Information Slightly soluble in water.
Formula Weight 246.07
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Pyrene-1-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only