missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolone Orange, TCI America™

Supplier:  TCI America P059625G

Catalog No. P059625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pyrazolone Orange
C32H24Cl2N8O2
25 g
HKYYDLYCGBDWSB-UHFFFAOYSA-N
4-[[2-chloro-4-[3-chloro-4-[(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)diazenyl]phenyl]phenyl]diazenyl]-5-methyl-2-phenyl-4H-pyrazol-3-one
20843803
Crystalline Powder
3520-72-7
MFCD00059727
Pigment Orange 13
CC1=NN(C(=O)C1N=NC2=C(C=C(C=C2)C3=CC(=C(C=C3)N=NC4C(=NN(C4=O)C5=CC=CC=C5)C)Cl)Cl)C6=CC=CC=C6
623.498
623.50

Chemical Identifiers

3520-72-7
623.498
HKYYDLYCGBDWSB-UHFFFAOYSA-N
20843803
CC1=NN(C(=O)C1N=NC2=C(C=C(C=C2)C3=CC(=C(C=C3)N=NC4C(=NN(C4=O)C5=CC=CC=C5)C)Cl)Cl)C6=CC=CC=C6
C32H24Cl2N8O2
MFCD00059727
Pigment Orange 13
4-[[2-chloro-4-[3-chloro-4-[(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)diazenyl]phenyl]phenyl]diazenyl]-5-methyl-2-phenyl-4H-pyrazol-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3193

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions