accessibility menu, dialog, popup

UserName

CAS RN 3520-72-7

IUPAC Name: 4-[2-(3,3'-dichloro-4'-{2-[(4E)-3-methyl-5-oxo-1-phenyl-4,5-dihydro-1H-pyrazol-4-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 4-[2-(3,3'-dichloro-4'-{2-[(4E)-3-methyl-5-oxo-1-phenylpyrazol-4-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-methyl-2-phenylpyrazol-3-one
Molecular Weight (g/mol): 623.5
Molecular Formula: C32H24Cl2N8O2
InChi Key: YFIMMXDLMXFWGF-IDVKPAOMSA-N
SMILES: CC1=NN(C(=O)C1=NNC1=CC=C(C=C1Cl)C1=CC=C(N\N=C2/C(C)=NN(C2=O)C2=CC=CC=C2)C(Cl)=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results