missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazolo[3,4-b]pyridine 98.0+%, TCI America™
Supplier: TCI America P22851G
Specifications
| Pyrazolo[3,4-b]pyridine | |
| 98°C | |
| 120°C | |
| MFCD05663981 | |
| 1h-pyrazolo 3,4-b pyridine, 2h-pyrazolo 3,4-b pyridine, 7-azaindazole, pyrazolo 3,4-b pyridine, 7-aza-1h-indazole, pyrazolo 5,4-b pyridine, 7-aza indazole, 7-azabenzpyrazole, acmc-1cs3b, 2h pyrazolo 3,4 b pyridin | |
| C1=CC2=C(NN=C2)N=C1 | |
| 119.127 | |
| 119.13 | |
| Crystalline Powder |
| 271-73-8 | |
| Yellow | |
| C6H5N3 | |
| 1 g | |
| GVLRTOYGRNLSDW-UHFFFAOYSA-N | |
| 1H-pyrazolo[3,4-b]pyridine | |
| 2755850 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 271-73-8 | |
| 119.127 | |
| GVLRTOYGRNLSDW-UHFFFAOYSA-N | |
| 2755850 | |
| C1=CC2=C(NN=C2)N=C1 |
| C6H5N3 | |
| MFCD05663981 | |
| 1h-pyrazolo 3,4-b pyridine, 2h-pyrazolo 3,4-b pyridine, 7-azaindazole, pyrazolo 3,4-b pyridine, 7-aza-1h-indazole, pyrazolo 5,4-b pyridine, 7-aza indazole, 7-azabenzpyrazole, acmc-1cs3b, 2h pyrazolo 3,4 b pyridin | |
| 1H-pyrazolo[3,4-b]pyridine |
Safety and Handling
TSCA : No