missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[3,4-b]pyridine 98.0+%, TCI America™

Catalog No. p-7135493
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2285200MG 200 mg
P22851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2285200MG Supplier TCI America Supplier No. P2285200MG
Only null left

Chemical Identifiers

CAS 271-73-8
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD05663981
InChI Key GVLRTOYGRNLSDW-UHFFFAOYSA-N
Synonym 1h-pyrazolo 3,4-b pyridine, 2h-pyrazolo 3,4-b pyridine, 7-azaindazole, pyrazolo 3,4-b pyridine, 7-aza-1h-indazole, pyrazolo 5,4-b pyridine, 7-aza indazole, 7-azabenzpyrazole, acmc-1cs3b, 2h pyrazolo 3,4 b pyridin
PubChem CID 2755850
IUPAC Name 1H-pyrazolo[3,4-b]pyridine
SMILES C1=CC2=C(NN=C2)N=C1

Specifications

Melting Point 98°C
Color Yellow
Boiling Point 120°C
Quantity 200 mg
Formula Weight 119.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazolo[3,4-b]pyridine