missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-2-carboxylic acid, Tech., Thermo Scientific™

$257.79 - $257.79

Chemical Identifiers

CAS 63237-88-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD06739057
InChI Key AYGNAOBAPQNTIL-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine-2-carboxylic acid, 2-carboxypyrazolo 1,5-a pyridine, h-pyrazolo 1,5-a pyridine-2-carboxylic acid, pubchem15887, 2-pyrazolo 1,5-a pyridinecarboxylic acid, 8-hydropyrazolo 1,5-a pyridine-2-carboxylic acid, pyrazolo 1 pound not5-a pyridine-2-carboxylicacid
PubChem CID 7127820
IUPAC Name pyrazolo[1,5-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=NN2C=CC=CC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC39401DA
SDS View Documents
Thermo Scientific Chemicals
CC39401DA
1 g Amber Glass Bottle
Each for $257.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

63237-88-7
162.15
AYGNAOBAPQNTIL-UHFFFAOYSA-N
7127820
OC(=O)C1=NN2C=CC=CC2=C1
C8H6N2O2
MFCD06739057
pyrazolo 1,5-a pyridine-2-carboxylic acid, 2-carboxypyrazolo 1,5-a pyridine, h-pyrazolo 1,5-a pyridine-2-carboxylic acid, pubchem15887, 2-pyrazolo 1,5-a pyridinecarboxylic acid, 8-hydropyrazolo 1,5-a pyridine-2-carboxylic acid, pyrazolo 1 pound not5-a pyridine-2-carboxylicacid
pyrazolo[1,5-a]pyridine-2-carboxylic acid

Specifications

63237-88-7
100.0
Amber Glass Bottle
MFCD06739057
pyrazolo 1,5-a pyridine-2-carboxylic acid, 2-carboxypyrazolo 1,5-a pyridine, h-pyrazolo 1,5-a pyridine-2-carboxylic acid, pubchem15887, 2-pyrazolo 1,5-a pyridinecarboxylic acid, 8-hydropyrazolo 1,5-a pyridine-2-carboxylic acid, pyrazolo 1 pound not5-a pyridine-2-carboxylicacid
OC(=O)C1=NN2C=CC=CC2=C1
162.15
162.15
Technical
96.5
97%
C8H6N2O2
1 g
AYGNAOBAPQNTIL-UHFFFAOYSA-N
pyrazolo[1,5-a]pyridine-2-carboxylic acid
7127820
97%
Pyrazolo[1, 5-a]pyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'