accessibility menu, dialog, popup

UserName

CAS RN 63237-88-7

IUPAC Name: pyrazolo[1,5-a]pyridine-2-carboxylic acid
Synonyms: pyrazolo[1,5-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: AYGNAOBAPQNTIL-UHFFFAOYSA-N
SMILES: OC(=O)C1=NN2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results