missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-2-carboxylique, Tech., Thermo Scientific™

Numéro de catalogue. CC39401DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC39401DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC39401DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC39401DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 63237-88-7
Formule moléculaire C8H6N2O2
Poids moléculaire (g/mol) 162.15
Numéro MDL MFCD06739057
Clé InChI AYGNAOBAPQNTIL-UHFFFAOYSA-N
Synonyme pyrazolo 1,5-a pyridine-2-carboxylic acid, 2-carboxypyrazolo 1,5-a pyridine, h-pyrazolo 1,5-a pyridine-2-carboxylic acid, pubchem15887, 2-pyrazolo 1,5-a pyridinecarboxylic acid, 8-hydropyrazolo 1,5-a pyridine-2-carboxylic acid, pyrazolo 1 pound not5-a pyridine-2-carboxylicacid
PubChem CID 7127820
Nom de l’IUPAC Acide pyrazolo[1,5-a]pyridine-2-carboxylique
SOURIRES OC(=O)C1=NN2C=CC=CC2=C1

Spécifications

Nom ou substance chimique Pyrazolo[1, 5-a]pyridine-2-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 162.15
Pourcentage de pureté 97%
Niveau Caractéristiques
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.