missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazole-3,5-dicarboxylic Acid Monohydrate 98.0+%, TCI America™
$186.41 - $529.40
Chemical Identifiers
| CAS | 3112-31-0 |
|---|---|
| Molecular Formula | C5H4N2O4 |
| Molecular Weight (g/mol) | 156.097 |
| MDL Number | MFCD00005235 |
| InChI Key | YDMVPJZBYSWOOP-UHFFFAOYSA-N |
| Synonym | 3,5-pyrazoledicarboxylic acid, pyrazole-3,5-dicarboxylic acid, pyrazole-3,5-dicarboxylic acid monohydrate, 3,5-pyrazol dicarboxylic acid, 3,5-pyrazole dicarboxylic acid, zlchem 726, 3,5-dicarboxypyrazole, acmc-20a0hc, ksc223c6j, 3,5-pyrazole-dicarboxylic acid |
| PubChem CID | 76559 |
| IUPAC Name | 1H-pyrazole-3,5-dicarboxylic acid |
| SMILES | C1=C(NN=C1C(=O)O)C(=O)O |
Chemical Identifiers
| 3112-31-0 | |
| 156.097 | |
| YDMVPJZBYSWOOP-UHFFFAOYSA-N | |
| 76559 | |
| C1=C(NN=C1C(=O)O)C(=O)O |
| C5H4N2O4 | |
| MFCD00005235 | |
| 3,5-pyrazoledicarboxylic acid, pyrazole-3,5-dicarboxylic acid, pyrazole-3,5-dicarboxylic acid monohydrate, 3,5-pyrazol dicarboxylic acid, 3,5-pyrazole dicarboxylic acid, zlchem 726, 3,5-dicarboxypyrazole, acmc-20a0hc, ksc223c6j, 3,5-pyrazole-dicarboxylic acid | |
| 1H-pyrazole-3,5-dicarboxylic acid |
Specifications
| 3112-31-0 | |
| White | |
| MFCD00005235 | |
| 3,5-pyrazoledicarboxylic acid, pyrazole-3,5-dicarboxylic acid, pyrazole-3,5-dicarboxylic acid monohydrate, 3,5-pyrazol dicarboxylic acid, 3,5-pyrazole dicarboxylic acid, zlchem 726, 3,5-dicarboxypyrazole, acmc-20a0hc, ksc223c6j, 3,5-pyrazole-dicarboxylic acid | |
| C1=C(NN=C1C(=O)O)C(=O)O | |
| 156.097 | |
| 156.10 | |
| Crystalline Powder |
| 295°C | |
| C5H4N2O4 | |
| 5 g | |
| YDMVPJZBYSWOOP-UHFFFAOYSA-N | |
| 1H-pyrazole-3,5-dicarboxylic acid | |
| 76559 | |
| ≥98.0% (T) | |
| Pyrazole-3,5-dicarboxylic Acid Monohydrate |
Safety and Handling
TSCA : No