accessibility menu, dialog, popup

UserName

OONHNOO

CAS RN 3112-31-0

IUPAC Name: 1H-pyrazole-3,5-dicarboxylate
Synonyms: 1H-pyrazole-3,5-dicarboxylate
Molecular Weight (g/mol): 154.08
Molecular Formula: C5H2N2O4
InChi Key: YDMVPJZBYSWOOP-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC(=NN1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results