missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazole-3,5-dicarboxylic Acid Monohydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 295°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 156.10 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyrazole-3,5-dicarboxylic Acid Monohydrate |
Safety and Handling
| TSCA | No |