missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinecarboxylic Acid 98.0+%, TCI America™

Catalog No. P094025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P094025G 25 g
P0940500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P094025G Supplier TCI America Supplier No. P094025G

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 124.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazinecarboxylic Acid
EINECSNumber (5)-0952
TSCA No