accessibility menu, dialog, popup

UserName

HOONN

CAS RN 98-97-5

IUPAC Name: pyrazine-2-carboxylic acid
Synonyms: pyrazinoic acid
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: NIPZZXUFJPQHNH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC=N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)