missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazine-2-carboxylic acid, 99%
CAS: 98-97-5 | C5H4N2O2 | 124.099 g/mol
Supplier: Thermo Scientific Chemicals A1336322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Pyrazine-2-carboxylic acid | |
| ∼225°C (decomposition) | |
| MFCD00006130 | |
| 112305 | |
| 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid | |
| C1=CN=C(C=N1)C(=O)O | |
| 124.099 | |
| CHEBI:71311 | |
| 99% |
| 98-97-5 | |
| C5H4N2O2 | |
| 100 g | |
| 14,7959 | |
| NIPZZXUFJPQHNH-UHFFFAOYSA-N | |
| pyrazine-2-carboxylic acid | |
| 1047 | |
| 124.1 |
Chemical Identifiers
| 98-97-5 | |
| 124.099 | |
| NIPZZXUFJPQHNH-UHFFFAOYSA-N | |
| 1047 | |
| pyrazine-2-carboxylic acid |
| C5H4N2O2 | |
| MFCD00006130 | |
| 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid | |
| CHEBI:71311 | |
| C1=CN=C(C=N1)C(=O)O |
Safety and Handling
EINECSNumber : 202-718-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only