missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carboxylic acid, 99%

Catalog No. AAA1336314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1336314 25 g
AAA1336322 100 g
AAA1336336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1336314 Supplier Thermo Scientific Chemicals Supplier No. A1336314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-97-5 | C5H4N2O2 | 124.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 25 g
Beilstein 112305
Merck Index 14,7959
Formula Weight 124.1
Percent Purity 99%
Chemical Name or Material Pyrazine-2-carboxylic acid
EINECSNumber 202-718-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only