missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2,5-dicarboxylic acid, 95%

Catalog No. p-7056053
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64795MD 250 mg
AAH6479503 1 g
AAH6479506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64795MD Supplier Thermo Scientific Chemicals Supplier No. H64795MD
Only null left

CAS: 122-05-4 | C6H4N2O4 | 168.11 g/mol

It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 122-05-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD00216919
InChI Key GMIOYJQLNFNGPR-UHFFFAOYSA-N
Synonym 2,5-pyrazinedicarboxylic acid, 2,5-pyrazinedicarboxylicacid, pubchem8868, 2,5-pyrazine-diacid, ksc177q1r, pyrazinedicarboxylic-2,5 acid
PubChem CID 255280
IUPAC Name pyrazine-2,5-dicarboxylic acid
SMILES OC(=O)C1=CN=C(C=N1)C(O)=O

Specifications

Melting Point 255°C to 260°C (literature)
Quantity 250 mg
Beilstein 139985
Solubility Information Slightly soluble in water.
Formula Weight 168.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrazine-2,5-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only