CAS RN 122-05-4
IUPAC Name:
pyrazine-2,5-dicarboxylic acid
Synonyms:
pyrazine-2,5-dicarboxylic acid
Molecular Weight (g/mol):
168.11
Molecular Formula:
C6H4N2O4
InChi Key:
GMIOYJQLNFNGPR-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN=C(C=N1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Pyrazine-2,5-dicarboxylic acid, 95%
Pyrazine-2,5-dicarboxylic acid, 95%
CAS: 122-05-4 Molecular Formula: C6H4N2O4 Molecular Weight (g/mol): 168.11 MDL Number: MFCD00216919 InChI Key: GMIOYJQLNFNGPR-UHFFFAOYSA-N Synonym: 2,5-pyrazinedicarboxylic acid,2,5-pyrazinedicarboxylicacid,pubchem8868,2,5-pyrazine-diacid,ksc177q1r,pyrazinedicarboxylic-2,5 acid PubChem CID: 255280 IUPAC Name: pyrazine-2,5-dicarboxylic acid SMILES: OC(=O)C1=CN=C(C=N1)C(O)=O