missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazinamide, 98%
CAS: 98-96-4 | C5H5N3O | 123.115 g/mol
$82.81 - $319.52
Chemical Identifiers
| CAS | 98-96-4 |
|---|---|
| Molecular Formula | C5H5N3O |
| Molecular Weight (g/mol) | 123.115 |
| MDL Number | MFCD00006132 |
| InChI Key | IPEHBUMCGVEMRF-UHFFFAOYSA-N |
| PubChem CID | 1046 |
| ChEBI | CHEBI:45285 |
| IUPAC Name | pyrazine-2-carboxamide |
| SMILES | C1=CN=C(C=N1)C(=O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6682209
|
Thermo Scientific Chemicals
J6682209 |
10 g |
N/A
|
|
|||||
|
AAJ6682214
|
Thermo Scientific Chemicals
J6682214 |
25 g |
N/A
|
|
|||||
|
AAJ6682222
|
Thermo Scientific Chemicals
J6682222 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-96-4 | |
| 123.115 | |
| IPEHBUMCGVEMRF-UHFFFAOYSA-N | |
| CHEBI:45285 | |
| C1=CN=C(C=N1)C(=O)N |
| C5H5N3O | |
| MFCD00006132 | |
| 1046 | |
| pyrazine-2-carboxamide |
Specifications
| Pyrazinamide | |
| 98-96-4 | |
| Odorless | |
| 10 g | |
| C5H5N3O | |
| 112306 | |
| Pyrazinoic acid amide; Pyrazinecarboxamide | |
| IPEHBUMCGVEMRF-UHFFFAOYSA-N | |
| pyrazine-2-carboxamide | |
| 1046 | |
| 123.11 | |
| Pyrazinamide |
| Powder | |
| 189°C to 191°C | |
| White | |
| >110°C (230°F) | |
| MFCD00006132 | |
| 14,7956 | |
| Soluble in water /n | |
| C1=CN=C(C=N1)C(=O)N | |
| 123.115 | |
| CHEBI:45285 | |
| 98% |
Safety and Handling
EINECSNumber : 202-717-6
RTECSNumber : UQ2275000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only