missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pyrantel pamoate, 98%
Supplier: Thermo Scientific™ 461891000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Pyrantel pamoate | |
| C11H14N2S·C23H16O6 | |
| Solubility in water: insoluble | |
| 594.68 | |
| CHEBI:8655 | |
| 98% |
| 22204-24-6 | |
| 100g | |
| AQXXZDYPVDOQEE-MXDQRGINSA-N | |
| 5281033 | |
| 594.68 |
Chemical Identifiers
| 22204-24-6 | |
| 594.68 | |
| 5281033 |
| C11H14N2S·C23H16O6 | |
| AQXXZDYPVDOQEE-MXDQRGINSA-N | |
| CHEBI:8655 |
Safety and Handling
EINECSNumber : 244-837-1
RUO â Research Use Only