missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pyrantel pamoate, 98%

Supplier:  Thermo Scientific™ 461890050

 View more versions of this product

Catalog No. AC461890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Pyrantel pamoate
C11H14N2S·C23H16O6
Solubility in water: insoluble
594.68
CHEBI:8655
98%
22204-24-6
5g
AQXXZDYPVDOQEE-MXDQRGINSA-N
5281033
594.68

Chemical Identifiers

22204-24-6
594.68
5281033
C11H14N2S·C23H16O6
AQXXZDYPVDOQEE-MXDQRGINSA-N
CHEBI:8655
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 244-837-1

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only