missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYGL Monoclonal Antibody (9#)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565977
Description
PYGL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a homodimeric protein that catalyses the cleavage of alpha-1,4-glucosidic bonds to release glucose-1-phosphate from liver glycogen stores. This protein switches from inactive phosphorylase B to active phosphorylase A by phosphorylation of serine residue 15. Activity of this enzyme is further regulated by multiple allosteric effectors and hormonal controls. Humans have three glycogen phosphorylase genes that encode distinct isozymes that are primarily expressed in liver, brain and muscle, respectively. The liver isozyme serves the glycemic demands of the body in general while the brain and muscle isozymes supply just those tissues. In glycogen storage disease type VI, also known as Hers disease, mutations in liver glycogen phosphorylase inhibit the conversion of glycogen to glucose and results in moderate hypoglycemia, mild ketosis, growth retardation and hepatomegaly. Alternative splicing results in multiple transcript variants encoding different isoforms.Chemical Identifiers
| PYGL | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| PYGL | |
| glycogen liver; Glycogen phosphorylase, liver form; GPLL; GSD6; Lgp; liver glycogen phosphorylase; phosphorylase, glycogen, liver; PYGL | |
| PYGL | |
| Primary | |
| Protein A/G |
| 9# | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06737, P09811, Q9ET01 | |
| 110095, 5836, 64035 | |
| Recombinant Glycogen Phosphorylase, Liver (PYGL) (Gln73-Val267). | |
| Antibody |
Specifications
| PYGL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06737, P09811, Q9ET01 | |
| PYGL | |
| Recombinant Glycogen Phosphorylase, Liver (PYGL) (Gln73-Val267). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9# | |
| Unconjugated | |
| PYGL | |
| glycogen liver; Glycogen phosphorylase, liver form; GPLL; GSD6; Lgp; liver glycogen phosphorylase; phosphorylase, glycogen, liver; PYGL | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 110095, 5836, 64035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |