missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PWP1 Recombinant Rabbit Monoclonal Antibody (23GB2580)

Catalog No. MA553557
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553557 20 μL
MA553556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553557 Supplier Invitrogen™ Supplier No. MA553557
Only null left

Rabbit Recombinant Monoclonal Antibody

PWP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen PWP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2580
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PWP1
Gene Accession No. Q13610, Q99LL5
Gene Alias 2310058A11Rik; 2610205J09Rik; AI098170; AL022879; AV083409; AW550690; C20orf152; endonuclein; IEF-SSP-9502; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; periodic tryptophan protein 1 homolog; Pwp1; PWP1 homolog (S. cerevisiae); PWP1 homolog, endonuclein; RGD1310705
Gene Symbols PWP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PWP1 (460-501AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103136, 11137
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less