missing translation for 'onlineSavingsMsg'
Learn More

Putrescine, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019830
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019830 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019830 Supplier Merck Emd Millipore Supplier No. 51799100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Putrescine is a naturally occurring polyamine, which is produced via breakdown of ornithine amino acid.

Specifications

CAS 110-60-1
Density 0.877 g/mL (at 25°C)
Physical Form Neat
Linear Formula NH2(CH2)4 NH2
MDL Number MFCD00008235
Melting Point 25°C to 28°C
Boiling Point 158°C to 160°C
Synonym 1,4-Diaminobutane; 1,4-Butanediamine; Putrescine; Tetramethylenediamine
Quantity 100 mg
Formula Weight 88.15
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less