missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUSL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556910 Supplier Invitrogen™ Supplier No. PA556910
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: WTLPADCLDM VAMQEAAQHL LGTHDFSAFQ SAGSPVPSPV RTLRRVSVSP GQASPLVTPE ESRKLRFWNL EFESQSFLY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

PUSL1 is a protein coding gene. Gene ontology (GO) annotation include intracellular membrane-bounded organelle.

Specifications

Antigen PUSL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PUSL1
Gene Accession No. Q8N0Z8
Gene Alias pseudouridylate synthase-like 1; PUSL1; tRNA pseudouridine synthase-like 1; tRNA pseudouridylate synthase-like 1; tRNA-uridine isomerase-like 1
Gene Symbols PUSL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PUSL1. Recombinant protein control fragment (Product #RP-94450).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less