Learn More
MilliporeSigma™ Supelco™ Push Button Valve
Catalog No. 111011728
Change view
Click to view available options
For Use With: Hamilton TLL Syringes
Catalog No.
111011728 For Use With: Hamilton TLL Syringes
1 options
Catalog No. 111011728 Supplier MilliporeSigma™ Supelco™ Supplier No. 22285
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
Specifications
Material PTFE (Mininert Valve)
Quantity 1/Pk.
Packaging 1/Pk.
For Use With (Equipment) Hamilton TLL Syringes