Learn More
PURUS™ 60-layer EnviroTack™ Contamination Control Mats
GREENER_CHOICE
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
Supplier:  PURUS™ PS 3645 64 G
 View more versions of this product
Catalog No. 19125836
Only null left
Description
Description
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
  • Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Certifications
Manufactured in ISO 9001 certified facility
Specifications
Chemical Identifiers
LDPE Blend
Specifications
36 x 45 in.
Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
60 Sheets/Mat, 4 Mats/Cs.
LDPE Blend
Film: 33 μm Adhesive: 8 μm
Regular Tack
Gray
Manufactured in an ISO 9001 certified facility
Product Suggestions
Product Suggestions