missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Purple Reagent Reservoir Chiller
Description
- Reagents are kept at less than 5°C (41°F) for approximately 3 hours
- Ice-free solution eliminates the risk of contamination from messy ice baths
- Stackable, save space when storing in the freezer or on the bench top
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
- Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
- Made from sonic welded, engineered polypropylene that ensures a leak free design
- Not intended for autoclave, do not use with liquid nitrogen
- Reusable
- Two-year warranty
Specifications
Specifications
| Capacity (English) | 3.4 oz. |
| Color | Purple |
| Capacity (Metric) | 100 mL |
| Dimensions (L x W x H) | 5.90 x 3.50 x 1.80 in. |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reservoir Cooler |
| Quantity | 2 Pk. |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Show More |