Learn More
Puromycin dihydrochloride, 98%
Puromycin dihydrochloride hydrate, CAS # 58-58-2, is an aminoglycoside showing antibiotic, anti-neoplastic, and anti-protozoal activities. This compound also has the ability to inhibit protein synthesis. | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol
Supplier: Thermo Scientific Chemicals J61278MC
| Quantity | 100 mg |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
General Information
• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis
Applications
• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.
Chemical Identifiers
| 58-58-2 | |
| 544.43 | |
| MKSVFGKWZLUTTO-MLYJQVMKNA-N | |
| Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1 |
| C22H31Cl2N7O5 | |
| MFCD00150080 | |
| 131632508 |
Specifications
| Puromycin Dihydrochloride | |
| Powder | |
| 168°C to 170°C | |
| 100 mg | |
| MFCD00150080 | |
| 14,7943 | |
| MKSVFGKWZLUTTO-MLYJQVMKNA-N | |
| (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride | |
| 131632508 | |
| 99% |
| Puromycin dihydrochloride | |
| 58-58-2 | |
| White | |
| C22H31Cl2N7O5 | |
| 3853613 | |
| Soluble in water | |
| Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1 | |
| 544.43 | |
| 544.44 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 200-387-8
RTECSNumber : AU355000
TSCA : No
Recommended Storage : Store at -20°C