missing translation for 'onlineSavingsMsg'
Learn More

Puromycin aminonucleoside, 98%

Catalog No. p-7076661
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66301MC 100 mg
AAJ66301ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66301MC Supplier Thermo Scientific Chemicals Supplier No. J66301MC
Only null left

Used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability. | CAS: 58-60-6 | C12H18N6O3 | 294.315 g/mol

Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Solubility
Soluble in water at 50mg/ml. May require heating

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 58-60-6
Molecular Formula C12H18N6O3
Molecular Weight (g/mol) 294.315
MDL Number MFCD00063462
InChI Key RYSMHWILUNYBFW-GRIPGOBMSA-N
Synonym 3'-Amino-3'-deoxy-N^6,N^6-dimethyladenosine
PubChem CID 6020
ChEBI CHEBI:42839
IUPAC Name (2R,3R,4S,5S)-4-amino-2-[6-(dimethylamino)purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
SMILES CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)N)O

Specifications

Type Puromycin Aminonucleoside
Physical Form Powder
Melting Point 215°C to 216°C
Packaging Plastic Bottle
Quantity 100 mg
Flash Point >110°C (230°F)
Beilstein 93902
Solubility Information Soluble in water at 50mg/ml. May require heating
Formula Weight 294.31
Percent Purity 98%
Chemical Name or Material Puromycin aminonucleoside
Show More Show Less
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 200-388-3
RTECSNumber AU7337000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only