accessibility menu, dialog, popup

UserName

CH3NH3CNNNNOOHH2NHO

CAS RN 58-60-6

IUPAC Name: 4-amino-2-[6-(dimethylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
Synonyms: puromycin aminonucleoside
Molecular Weight (g/mol): 294.32
Molecular Formula: C12H18N6O3
InChi Key: RYSMHWILUNYBFW-UHFFFAOYNA-N
SMILES: CN(C)C1=C2N=CN(C3OC(CO)C(N)C3O)C2=NC=N1
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results