missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puromycin aminonucleoside, 98%
Description
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.
Solubility
Soluble in water at 50mg/ml. May require heating
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.
Solubility
Soluble in water at 50mg/ml. May require heating
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Type | Puromycin Aminonucleoside |
| Physical Form | Powder |
| Melting Point | 215°C to 216°C |
| Packaging | Plastic Bottle |
| Quantity | 100 mg |
| Flash Point | >110°C (230°F) |
| Beilstein | 93902 |
| Solubility Information | Soluble in water at 50mg/ml. May require heating |
| Formula Weight | 294.31 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H373 |
| Precautionary Statement | P260-P314-P501c |
| EINECSNumber | 200-388-3 |
| RTECSNumber | AU7337000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only