missing translation for 'onlineSavingsMsg'
Learn More

Puritan™ PurFlock™ Ultra Flocked Swabs

Ideal for rapid diagnostic tests

Supplier:  Puritan™ 253706U

 View more versions of this product

Catalog No. 22-029-510

Only null left

Description

Description

Elongated Flock Tipped Specimen Collection Applicator with Plastic Handle

PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultra’s high purity characteristics makes it an ideal swab for various rapid diagnostic tests.

Compliance

FDA Good Manufcturing Practices (GMP)

Specifications

Specifications

CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
For collection of Microorganisms, Buccal cells and forensic evidence; molded breakpoint at 100 mm
152.4 mm
100 mm
Molded Breakpoint at 100 mm
5.5 mm
Individually-wrapped
Polystyrene
6 in.
Swab
4 in.
0.685 in.
17.39 mm
0.219 in.
Sterile
Elongated Flocked
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions