Learn More
Puritan™ UniTranz-RT Transport Systems
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Supplier:  Puritan™ UT361
 View more versions of this product
Catalog No. 22-027-110
Only null left
Explore available promotions
Description
Description
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA
Specifications
Specifications
UniTranz-RT Transport System
Sterile
CE
Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Individually-wrapped
300 Cs.
2°C to 25°C
Standard
Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Single Applicator
3 mL
Regular Polyester tip swab, wrapped and 3mL Universal Transport Medium Filled Vial
7.3 ±0.2
Plastic
Polyester
Product Suggestions
Product Suggestions