Learn More
Puritan™ HydraFlock™ Flocked Swabs
For collection and release of biological specimens
 View more versions of this product
Catalog No. 22025197
Only null left
Description
Description
A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.; For buccal cell collection; Specimen collection; Tip material is comprised of polyester bicomponent fiber; Made in USA
Specifications
Chemical Identifiers
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Sterile
Flocked
Swab
Polystyrene
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2 cm
15.2 cm
1.7 cm
0.2 in.
Polystyrene
Flocked
0.1 in.
For collection and release of biological specimens, buccal cell collection, specimen collection
0.254 cm
6 in.
0.69 in.
0.51 cm
Sterile
Standard
Product Suggestions
Product Suggestions