missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Purifier™ Logic™+ Class II B2 Biosafety Cabinets, US Models
Total exhaust biohazard cabinets with advanced ergonomic design that excels in user comfort and functionality.
Supplier: Labconco™ 303680001
Description
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile (CAP) airflow monitoring system
- Intrinsically-safe negative pressure design
- Air-Wave Entry System
- Contain-Air Negative Pressure Channel
- Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
- Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available.
- Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
- Fully-closing, clear tempered safety glass sash
- Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
- Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. One outlet on each side, with ground fault interruption (115 V)
- Smart-Start System user-programmable start up and shut down
- Leak-tight stainless steel interior and powder-coated steel exterior
- Removable towel catch located under work surface
- Code-activated electronic security lock
- 10° Angled sash with counterbalanced, easy lift mechanism
- ADA-Compliant touchpad control on right side post
- Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
- Heat load 990 BTU/hour (4 ft. models)
- Nominal inflow velocity of 105 fpm (0.5 m/sec)
- Nominal downflow velocity of 55 fpm (0.3 m/sec)
- 10 ft. (3 m) power cord with plug
- Five year warranty
- NSF International listed
- Optional NSF-approved non-welded telescoping base stand included on some models
- Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass Cord and Cable Portal) included on some models
Chemical Identifiers
| ADA, CAN/CSA C22.2, ETL, NSF 49, UL | |
| Type B2 Biosafety Cabinet | |
| <63 dBA | |
| North America (115 V, 15 A) | |
| 72.6 in. | |
| 184.4 cm | |
| 5 Years | |
| 79.25 cm |
| Stainless Steel | |
| Purifier Logic+ | |
| 4 | |
| 2 | |
| 20.3 cm | |
| 8 in. | |
| 31.2 in. | |
| 115 V, 60 Hz, 12 A |
Specifications
| ADA, CAN/CSA C22.2, ETL, NSF 49, UL | |
| 184.2 cm | |
| Purifier Logic+ | |
| 4 | |
| 290 W | |
| 72.6 in. | |
| 6.5 ft. | |
| 198.88 cm | |
| LED | |
| 8 in. | |
| 6 ft. | |
| 79.25 cm |
| Stainless Steel | |
| Type B2 Biosafety Cabinet | |
| <63 dBA | |
| North America (115 V, 15 A) | |
| 2 | |
| 20.3 cm | |
| 0.575 m3sec. | |
| Without Accessory Package | |
| 184.4 cm | |
| 5 Years | |
| 31.2 in. | |
| 115 V, 60 Hz, 12 A |