missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Logic™+ Class II B2 Biosafety Cabinets, International Models

Total exhaust biohazard cabinets with advanced ergonomic design that excels in user comfort and functionality.

Supplier:  Labconco™ 303680011

 View more versions of this product

Catalog No. 303680011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave Entry System
  • Contain-Air Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available.
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start System user-programmable start up and shut down
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Heat load 990 BTU/hour (4 ft. models)
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • 10 ft. (3 m) power cord with plug
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass Cord & Cable Portal) included on some models
  • Supporting base required if non-welded stand model is not selected
Specifications

Chemical Identifiers

Stainless Steel
Purifier Logic+
2
20.3 cm
8 in.
50/60 Hz
79.25 cm
Type B2 Biosafety Cabinet
<63 dBA
72.6 in.
184.4 cm
5 Years
31.2 in.

Specifications

ADA, CAN/CSA C22.2, CE, ETL, NSF 49, UL
184.2 cm
Purifier Logic+
2
290 W
72.6 in.
6.5 ft.
198.88 cm
LED
8 in.
6 ft.
31.2 in.
230 V, 50/60 Hz, 6 A
Stainless Steel
Type B2 Biosafety Cabinet
<63 dBA
North America (230 V)
2
20.3 cm
0.575 m3sec.
Without Accessory Package
184.4 cm
5 Years
50/60 Hz
79.25 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions