missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Logic™+ Class II B2 Biosafety Cabinets with Base Stand, US Models

Catalog No. 303480101
Change view
Click to view available options
Width (English) Interior:
4 ft.
6 ft.
Includes:
With Accessory Package
Without Accessory Package
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Interior Includes
303480101 4 ft. Without Accessory Package
303481101 4 ft. With Accessory Package
303680101 6 ft. Without Accessory Package
303681101 6 ft. With Accessory Package
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 303480101 Supplier Labconco™ Supplier No. 303480101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Total exhaust biohazard cabinets with advanced ergonomic design that excels in user comfort and functionality.

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave Entry System
  • Contain-Air Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available.
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start System user-programmable start up and shut down
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Heat load 990 BTU/hour (4 ft. models)
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • 10 ft. (3 m) power cord with plug
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass Cord and Cable Portal) included on some models

Specifications

Certifications/Compliance ADA, CAN/CSA C22.2, ETL, NSF 49, UL
Chamber Material Stainless Steel
Width (Metric) Interior 123.2 cm
Product Type Type B2 Biosafety Cabinet
Model Purifier Logic+
Noise Level <63 dBA
Outlets 4
Plug Type North America (115 V, 15 A)
Power Consumption 170 W
Receptacles 2
Height (English) 100.1 to 106.1 in.
Sash Opening (Metric) 20.3 cm
Width (English) Exterior 4.5 ft.
Volume (Metric) Exhaust 0.382 m3sec.
Width (Metric) Exterior 137.92 cm
Includes Without Accessory Package
Lighting LED
Height (Metric) 254.2 to 269.2 cm
Sash Opening (English) 8 in.
Warranty 5 Years
Width (English) Interior 4 ft.
Depth (English) Exterior 31.2 in.
Depth (Metric) Exterior 79.25 cm
Electrical Requirements 115 V, 60 Hz, 12 A
Show More Show Less