missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, US Models

Catalog No. 302610001
Change view
Click to view available options
Height (English):
89.2 to 95.2 in.
61.7 in.
Width (English):
3 ft.
4 ft.
5 ft.
6 ft.
Sash Opening (English):
8 in.
10 in.
Includes:
Cabinet
Cabinet, Accessory Package
Cabinet, Accessory Package, Base Stand
Cabinet, Base Stand
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English) Sash Opening (English) Includes
302610001 61.7 in. 6 ft. 10 in. Cabinet
302310001 61.7 in. 3 ft. 10 in. Cabinet
302310101 89.2 to 95.2 in. 3 ft. 10 in. Cabinet, Base Stand
302311001 61.7 in. 3 ft. 10 in. Cabinet, Accessory Package
302311101 89.2 to 95.2 in. 3 ft. 10 in. Cabinet, Accessory Package, Base Stand
302380001 61.7 in. 3 ft. 8 in. Cabinet
302380101 89.2 to 95.2 in. 3 ft. 8 in. Cabinet, Base Stand
302381001 61.7 in. 3 ft. 8 in. Cabinet, Accessory Package
302381101 89.2 to 95.2 in. 3 ft. 8 in. Cabinet, Accessory Package, Base Stand
302410001 61.7 in. 4 ft. 10 in. Cabinet
302410101 89.2 to 95.2 in. 4 ft. 10 in. Cabinet, Base Stand
302411001 61.7 in. 4 ft. 10 in. Cabinet, Accessory Package
302480001 61.7 in. 4 ft. 8 in. Cabinet
302480101 89.2 to 95.2 in. 4 ft. 8 in. Cabinet, Base Stand
302481001 61.7 in. 4 ft. 8 in. Cabinet, Accessory Package
302481101 89.2 to 95.2 in. 4 ft. 8 in. Cabinet, Accessory Package, Base Stand
302510001 61.7 in. 5 ft. 10 in. Cabinet
302511001 61.7 in. 5 ft. 10 in. Cabinet, Accessory Package
302511101 89.2 to 95.2 in. 5 ft. 10 in. Cabinet, Accessory Package, Base Stand
302580001 61.7 in. 5 ft. 8 in. Cabinet
302580101 89.2 to 95.2 in. 5 ft. 8 in. Cabinet, Base Stand
302581001 61.7 in. 5 ft. 8 in. Cabinet, Accessory Package
302581101 89.2 to 95.2 in. 5 ft. 8 in. Cabinet, Accessory Package, Base Stand
302610101 89.2 to 95.2 in. 6 ft. 10 in. Cabinet, Base Stand
302611001 61.7 in. 6 ft. 10 in. Cabinet, Accessory Package
302611101 89.2 to 95.2 in. 6 ft. 10 in. Cabinet, Accessory Package, Base Stand
302680001 61.7 in. 6 ft. 8 in. Cabinet
302680101 89.2 to 95.2 in. 6 ft. 8 in. Cabinet, Base Stand
302681001 61.7 in. 6 ft. 8 in. Cabinet, Accessory Package
302681101 89.2 to 95.2 in. 6 ft. 8 in. Cabinet, Accessory Package, Base Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Catalog No. 302610001 Supplier Labconco™ Supplier No. 302610001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models

Specifications

Certifications/Compliance NSF, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel
Width (Metric) Interior 184.2 cm
Product Type Type A2 Biosafety Cabinet
Model Purifier Logic+
Noise Level <63 dBA
Outlets 4
Plug Type North America (115 V, 20 A)
Power Consumption 570 W
Receptacles 2
Height (English) 61.7 in.
Sash Opening (Metric) 25.4 cm
Width (English) Exterior 78.3 in.
Volume (Metric) Exhaust 0.238 to 0.261 m3sec.
Width (Metric) Exterior 198.882 cm
Includes Cabinet
Lighting LED
Height (Metric) 156.718 cm
Sash Opening (English) 10 in.
Warranty 5 Years
Width (English) Interior 72.5 in.
Width (English) 6 ft.
Width (Metric) 198.882 cm
Frequency 60 Hz
Depth (English) Exterior 31.2 in.
Depth (Metric) Exterior 79.248 cm
Electrical Requirements 115 V, 60 Hz, 16 A
Voltage 115 V
Show More Show Less