missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, 6 ft. Width, International Models

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Supplier:  Labconco™ 302680111

 View more versions of this product

Catalog No. 302680111

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Specifications

Chemical Identifiers

Stainless Steel
Type A2 Biosafety Cabinet
<63 dBA
78.3 in.
5 Years
198.882 cm
31.2 in.
184.2 cm
Purifier Logic+
2
198.882 cm
72.5 in.
50/60 Hz
79.248 cm

Specifications

NSF, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2
184.2 cm
Purifier Logic+
2
550 W
89.2 to 95.2 in.
78.3 in.
198.882 cm
LED
8 in.
72.5 in.
50/60 Hz
79.248 cm
230 V
Stainless Steel
Type A2 Biosafety Cabinet
<63 dBA
North America (230 V)
2
20.3 cm
0.190 to 0.209 m3sec.
Cabinet, Base Stand
226.6 to 241.8 cm
5 Years
198.882 cm
31.2 in.
230 V, 50/60 Hz, 8 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions