missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, 5 ft. Width, International Models

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

$27,874.32 - $30,951.03

Specifications

Chamber Material Stainless Steel
Width (Metric) Interior 153.7 cm
Product Type Type A2 Biosafety Cabinet
Model Purifier Logic+
Noise Level <63 dBA
View More Specs

Products 8
Catalog Number Mfr. No. Plug Type Sash Opening (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Sash Opening (English) Includes Price Quantity & Availability  
302510011
View Documents Promotion Details
Labconco™
302510011
North America (230 V) 10 in. Cabinet N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302510111
View Documents Promotion Details
Labconco™
302510111
North America (230 V) 10 in. Cabinet, Base Stand N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302511011
View Documents Promotion Details
Labconco™
302511011
North America (230 V) 10 in. Cabinet, Accessory Package N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302511111
View Documents Promotion Details
Labconco™
302511111
North America (230 V) 10 in. Cabinet, Accessory Package, Base Stand N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302580011
View Documents Promotion Details
Labconco™
302580011
North America (230 V) 8 in. Cabinet N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302580111
View Documents Promotion Details
Labconco™
302580111
North America (230 V) 8 in. Cabinet, Base Stand N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302581011
View Documents Promotion Details
Labconco™
302581011
North America (230 V) 8 in. Cabinet, Accessory Package N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
302581111
View Documents Promotion Details
Labconco™
302581111
North America (230 V) 8 in. Cabinet, Accessory Package, Base Stand N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Specifications

Specifications

Stainless Steel
Type A2 Biosafety Cabinet
<63 dBA
66.3 in.
5 Years
168.402 cm
31.2 in.
153.7 cm
Purifier Logic+
2
168.402 cm
60.5 in.
50/60 Hz
79.248 cm
Videos
SDS
Documents

Documents