missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, 4 ft. Width, International Models
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
Supplier: Labconco™ 302411011
Description
All models feature:
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile (CAP) airflow monitoring system
- Intrinsically-safe negative pressure design
- Air-Wave™ Entry System
- Contain-Air™ Negative Pressure Channel
- Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
- Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
- Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
- Fully-closing, clear 1/4 in. tempered safety glass sash
- Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
- Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
- Smart-Start™ System user-programmable start up and shut down
- Night-Smart™ System reduces blower speed when sash is closed
- Leak-tight stainless steel interior and powder-coated steel exterior
- Removable towel catch located under work surface
- Code-activated electronic security lock
- 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
- 10° Angled sash with counterbalanced, easy lift mechanism
- ADA-Compliant touchpad control on right side post
- Noise level <63 dBA
- Heat load 990 BTU/hour (4 ft. models)
- Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
- Nominal inflow velocity of 105 fpm (0.5 m/sec)
- Nominal downflow velocity of 55 fpm (0.3 m/sec)
- Approximately 70% air recirculation
- Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
- 10 ft (3 m) power cord with plug
- Built-in timer and digital clock
- Five year warranty
- NSF International listed
- Optional NSF-approved non-welded telescoping base stand included on some models
- Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Chemical Identifiers
| Stainless Steel | |
| Type A2 Biosafety Cabinet | |
| <63 dBA | |
| 54.3 in. | |
| 5 Years | |
| 137.922 cm | |
| 31.2 in. |
| 123.2 cm | |
| Purifier Logic+ | |
| 2 | |
| 137.922 cm | |
| 48.5 in. | |
| 50/60 Hz | |
| 79.248 cm |
Specifications
| NSF, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
| 123.2 cm | |
| Purifier Logic+ | |
| 2 | |
| 280 W | |
| 61.7 in. | |
| 54.3 in. | |
| 137.922 cm | |
| LED, UV | |
| 10 in. | |
| 5 Years | |
| 137.922 cm | |
| 31.2 in. | |
| 230 V, 50/60 Hz, 6 A |
| Stainless Steel | |
| Type A2 Biosafety Cabinet | |
| <63 dBA | |
| North America (230 V) | |
| 2 | |
| 25.4 cm | |
| 0.159 to 0.175 m3sec. | |
| Cabinet, Accessory Package | |
| 156.718 cm | |
| One | |
| 48.5 in. | |
| 50/60 Hz | |
| 79.248 cm | |
| 230 V |