missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Purifier™ Cell Logic™+ Class II B2 Biosafety Cabinets with Temp-Zone and Base Stand
Total exhaust B2 biohazard cabinets with advanced ergonomic design and temperature-controlled surface excel in user comfort and functionality
Supplier: Labconco™ 323481111
Description
All models feature:
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile (CAP) airflow monitoring system
- Intrinsically-safe negative pressure design
- Air-Wave Entry System
- Contain-Air Negative Pressure Channel
- Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
- Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available
- Bright, 90 to 150 footcandle, glare-free LED lighting located outside the contaminated work area
- Fully-closing, clear 1/4 in. tempered safety glass sash
- Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
- Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
- Smart-Start™ System user-programmable start up and shut down
- Leak-tight stainless steel interior and powder-coated steel exterior
- Removable towel catch located under work surface
- Code-activated electronic security lock
- 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
- 10° Angled sash with counterbalanced, easy lift mechanism
- ADA-Compliant touchpad control on right side post
- Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. (27 cm) wide x 12.0 in. (30 cm) deep (7.0 x 12 in. on 3 ft. models) located on the left side of the work surface. Circulator is required (not included). Circulator requirements: minimum flow rate of 0.4 to 0.5 GPM (1.5 to 1.9 LPM), typical volume of 6 liters, and temperature of 0 to 50° C (chiller/heater)
- Noise level <63 dBA
- Heat load 990 BTU/hour (4 ft. models)
- Nominal inflow velocity of 105 fpm (0.5 m/sec)
- Nominal downflow velocity of 55 fpm (0.3 m/sec)
- 10 ft. (3 m) power cord with plug
- Five year warranty
- NSF International compliant
- Optional NSF-approved non-welded telescoping base stand included on some models
- Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord and Cable Portal) included on some models
- All models require supporting base if non-welded stand option is not selected
Chemical Identifiers
| Stainless Steel | |
| Purifier Cell Logic+ | |
| 2 | |
| 20.3 cm | |
| LED, UV | |
| 8 in. | |
| 31.2 in. |
| Type B2 Biosafety Cabinet | |
| <63 dBA | |
| 100.1 to 106.1 in. | |
| Cabinet with Accessory and Temp-zone Packages, Base Stand | |
| 254.2 to 269.2 cm | |
| 5 Years | |
| 79.248 cm |
Specifications
| NSF, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
| 123.2 cm | |
| Purifier Cell Logic+ | |
| 2 | |
| 170 W | |
| 100.1 to 106.1 in. | |
| 54.3 in. | |
| 137.922 cm | |
| LED, UV | |
| 8 in. | |
| 5 Years | |
| 137.922 cm | |
| 31.2 in. | |
| 230 V, 50/60 Hz, 6 A |
| Stainless Steel | |
| Type B2 Biosafety Cabinet | |
| <63 dBA | |
| North America (230 V) | |
| 2 | |
| 20.3 cm | |
| 0.382 m3sec. | |
| Cabinet with Accessory and Temp-zone Packages, Base Stand | |
| 254.2 to 269.2 cm | |
| One | |
| 4 ft. | |
| 50/60 Hz | |
| 79.248 cm | |
| 230 V |