missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Cell Logic™+ Class II A2 Biosafety Cabinets with Temp-Zone, US Models

Catalog No. 322491001
Change view
Click to view available options
Width (English) Interior:
3 ft.
4 ft.
5 ft.
6 ft.
Includes:
Accessory and Temp-zone Packages
Accessory and Temp-zone Packages, Base Stand
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Interior Includes
322491001 4 ft. Accessory and Temp-zone Packages
322391001 3 ft. Accessory and Temp-zone Packages
322391101 3 ft. Accessory and Temp-zone Packages, Base Stand
322491101 4 ft. Accessory and Temp-zone Packages, Base Stand
322591001 5 ft. Accessory and Temp-zone Packages
322591101 5 ft. Accessory and Temp-zone Packages, Base Stand
322691001 6 ft. Accessory and Temp-zone Packages
322691101 6 ft. Accessory and Temp-zone Packages, Base Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 322491001 Supplier Labconco™ Supplier No. 322491001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biohazard cabinets with advanced ergonomic design and temperature-controlled surface excel in user comfort and functionality

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90 to 150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. (27 cm) wide x 12.0 in. (30 cm) deep (7 x 12 in. on 3 ft. models) located on the left side of the work surface. Circulator is required (not included). Circulator requirements: minimum flow rate of 0.4 to 0.5 GPM (1.5 to 1.9 LPM), typical volume of 6 liters, and temperature of 0 to 50° C (chiller/heater)
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft. (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International compliant
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord and Cable Portal) included on some models
  • All models require supporting base if non-welded stand option is not selected

Specifications

Certifications/Compliance NSF, Modified ASHRAE 110, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel
Width (Metric) Interior 123.2 cm
Product Type Type A2 Biosafety Cabinet
Model Purifier Cell Logic+
Noise Level <63 dBA
Outlets 4
Plug Type North America (115 V, 15 A)
Power Consumption 275 W
Receptacles 2
Height (English) 61.7 in.
Sash Opening (Metric) 22.9 cm
Width (English) Exterior 54.3 in.
Volume (Metric) Exhaust 0.15 m3sec.
Width (Metric) Exterior 156.718 cm
Includes Accessory and Temp-zone Packages
Lighting LED, UV
Height (Metric) 156.718 cm
Sash Opening (English) 9 in.
Service Fixtures One
Warranty 5 Years
Width (English) Interior 4 ft.
Width (Metric) 137.922 cm
Frequency 60 Hz
Depth (English) Exterior 31.2 in.
Depth (Metric) Exterior 79.248 cm
Electrical Requirements 115 V, 60 Hz, 12 A
Voltage 115 V
Show More Show Less