missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Cell Logic™+ Class II A2 Biosafety Cabinets with Scope-Ready and Temp-Zone with Base Stand, International Models

Catalog No. p-7160677
Change view
Click to view available options
Plug Type:
North America (230 V)
Width (English) Interior:
3 ft.
4 ft.
5 ft.
6 ft.
Outlets:
2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Width (English) Interior Outlets
342391111 North America (230 V) 3 ft. 2
342491111 North America (230 V) 4 ft. 2
342591111 North America (230 V) 5 ft. 2
342691111 North America (230 V) 6 ft. 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 342391111 Supplier Labconco™ Supplier No. 342391111

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Microscope-ready biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90 to 150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior & powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening & 27.0 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar. Stand-Still™ Isolation Platform is required and sold separately
  • Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. (27 cm) wide x 12.0 in (30 cm) deep (7.0 x 12.0 in. on 3 ft. models) located on the left side of the work surface. Circulator is required (not included). Circulator requirements: minimum flow rate of 0.4-0.5 GPM (1.5-1.9 LPM), typical volume of 6 liters, and temperature of 0 to 50° C (chiller/heater)
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft. (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International compliant
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (ADA-compliant service fixture(s) with ball-type valve(s) and NSF-Approved Vacu-Pass™ Cord and Cable Portal) included on some models
  • All models require supporting base if non-welded stand option is not selected

Specifications

Certifications/Compliance NSF, Modified ASHRAE 110, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel
Width (Metric) Interior 92.7 cm
Product Type Type A2 Biosafety Cabinet
Model Purifier Cell Logic+
Noise Level <63 dBA
Outlets 2
Plug Type North America (230 V)
Power Consumption 230 W
Receptacles 2
Height (English) 89.3 to 95.3 in.
Sash Opening (Metric) 22.9 cm
Width (English) Exterior 42.3 in.
Volume (Metric) Exhaust 0.113 m3sec.
Width (Metric) Exterior 226.8 to 242.1 cm
Includes Cabinet with Accessory, Temp-zone and Scope-ready Packages, Base Stand
Lighting LED
Height (Metric) 226.8 to 242.1 cm
Sash Opening (English) 9 in.
Service Fixtures One
Warranty 5 Years
Width (English) Interior 3 ft.
Width (Metric) 107.442 cm
Frequency 50/60 Hz
Depth (English) Exterior 31.2 in.
Depth (Metric) Exterior 79.248 cm
Electrical Requirements 230 V, 50/60 Hz, 6 A
Voltage 230 V
Show More Show Less