missing translation for 'onlineSavingsMsg'
Learn More

Purified Water, USP, Cytiva Hyclone™

Catalog No. WPWUSN1X6
Change view
Click to view available options
Quantity:
20 L
6 x 1 L
200 L
Packaging:
Cubitainer
Drum
PET Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
WPWUSN1X6 6 x 1 L PET Plastic Bottle
WPWUSN20L 20 L Cubitainer
WPWUSN2XL 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. WPWUSN1X6 Supplier Cytiva Supplier No. WPWUSN1X6
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Purified Water, USP can be used as an excipient in the production of non-parenteral preparations such as cleaning of certain equipment and non-parenteral product-contact components.

  • Complies with the USP monograph for sterile purified water
  • Available in sterile and non-sterile formats
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Purified utilizing a nine-step process resulting in the highest quality water on the market
  • Distilled
  • Deionized

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Conductivity Meets Current USP Requirement
Grade USP
Packaging PET Plastic Bottle
Sterility Non-sterile
Certificate of Analysis Yes
Quantity 6 x 1 L
ShelfLife 36 Months
Recommended Storage 2-30°C